Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD Antibody (IgD26), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP250437R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250437R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250437R Supplier Novus Biologicals Supplier No. NBP250437R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgD Monoclonal specifically detects IgD in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen IgD
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone IgD26
Conjugate DyLight 550
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Gene Symbols IGHD
Host Species Mouse
Immunogen IgD purified from human myeloma serum
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3495
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.