Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD Antibody (IGHD/8367R), Novus Biologicals™
SDP

Catalog No. NB232564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB232564 100 μg
NB233275 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB232564 Supplier Novus Biologicals Supplier No. NBP320495100UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen IgD
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IGHD/8367R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 1mM PBS with 0.05% BSA
Gene Alias IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Host Species Rabbit
Immunogen Recombinant fragment (around aa1-200) of human IgD protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3495
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.