Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD Antibody (V88P3F7*D2), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20438A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20438A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20438A94 Supplier Novus Biologicals Supplier No. NBP250438AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgD Monoclonal specifically detects IgD in Fish samples. It is validated for ELISA.

Specifications

Antigen IgD
Applications ELISA
Classification Monoclonal
Clone V88P3F7*D2
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Gene Symbols IGHD
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3495.0
Target Species Fish
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.