Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGDCC3 Antibody (920038), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB310722 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310722 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310722 Supplier Novus Biologicals Supplier No. FAB8559PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGDCC3 Monoclonal specifically detects IGDCC3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IGDCC3
Applications Flow Cytometry
Classification Monoclonal
Clone 920038
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias immunoglobulin superfamily DCC subclass member 3, immunoglobulin superfamily, DCC subclass, member 3, Putative neuronal cell adhesion moleculePUNCHsT18880
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human IGDCC3, Met1-Gly640, Accession # Q81VU1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9543
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.