Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgE Antibody (4G7), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB359262 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359262 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359262 Supplier Novus Biologicals Supplier No. NB500471JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgE Monoclonal antibody specifically detects IgE in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen IgE
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 4G7
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias IgE, immunoglobulin heavy constant epsilon
Host Species Mouse
Immunogen Purified human IgE
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3497
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.