Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-I/IGF-1 Antibody (IGF1/2872R), HRP, Novus Biologicals™
SDP

Catalog No. NB380688 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380688 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380688 Supplier Novus Biologicals Supplier No. NBP308416H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IGF-I/IGF-1 Monoclonal antibody specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen IGF-I/IGF-1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone IGF1/2872R
Conjugate HRP
Formulation PBS
Gene Alias IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C
Host Species Rabbit
Immunogen Recombinant full-length human IGF-I/IGF-1 protein (Uniprot: P05019)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3479
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.