Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-I/IGF-1 Antibody (SPM406), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB005900 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB005900 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB005900 Supplier Novus Biologicals Supplier No. NBP234409AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGF-I/IGF-1 Monoclonal specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen IGF-I/IGF-1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM406
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C
Gene Symbols IGF1
Host Species Mouse
Immunogen Purified human IGF-I/IGF-1 protein (Uniprot: P05019)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Test Specificity This antibody is specific to Insulin-like Growth Factor (IGF-1) and shows minimal cross-reaction with IGF-11, Proinsulin, MSF, and Insulin. IGF-1 is a polypeptide growth factor with two isoforms that are produced by alternative splicing. Isoform 1 is also known as IGF-IB while isoform 2 is known as IGF-IA. IGF-1 stimulates the proliferation of a wide range of cell types including muscle, bone and cartilage tissue. It functions as an autocrine regulator of growth. Activation of IGF system has emerged as a key factor for tumor progression and resistance to apoptosis in many cancers like those of breast, thyroid and colon.
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.