Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-I Mouse, Clone: M23, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | IGF-I |
---|---|
Clone | M23 |
Dilution | Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, SDS-Page |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP234249F
![]() |
Novus Biologicals
NBP2342490.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP234249A
![]() |
Novus Biologicals
NBP2342490.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP234249B
![]() |
Novus Biologicals
NBP2342490.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
IGF-I/IGF-1 Monoclonal specifically detects IGF-I/IGF-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IGF-I | |
Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Rabbit | |
P05019 | |
3479 | |
Purified human IGF-1 protein | |
Primary | |
Store at 4C. | |
7.6 kDa |
M23 | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C | |
IGF1 | |
IgG1 κ | |
Protein A purified | |
This antibody is specific to Insulin-like Growth Factor (IGF-1) and shows minimal cross-reaction with IGF-11, Proinsulin, MSF, and Insulin. IGF-1 is a polypeptide growth factor with two isoforms that are produced by alternative splicing. Isoform 1 is also known as IGF-IB while isoform 2 is known as IGF-IA. IGF-1 stimulates the proliferation of a wide range of cell types including muscle, bone and cartilage tissue. It functions as an autocrine regulator of growth. Activation of IGF system has emerged as a key factor for tumor progression and resistance to apoptosis in many cancers like those of breast, thyroid and colon. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title