Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-I R/IGF1R Antibody (17-69), Novus Biologicals™
SDP

Catalog No. NB500705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500705 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500705 Supplier Novus Biologicals Supplier No. NB500705

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IGF-I R/IGF1R Monoclonal specifically detects IGF-I R/IGF1R in Human, Porcine, Rabbit samples. It is validated for Immunoprecipitation, Mass Spectrometry, Inhibition Activity.

Specifications

Antigen IGF-I R
Applications Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone 17-69
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 ug/ml, Mass Spectrometry, Inhibition Activity
Gene Accession No. P08069
Gene Alias CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
Gene Symbols IGF1R
Host Species Mouse
Immunogen IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Purification Method Protein G purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3480
Target Species Human, Pig, Rabbit
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.