Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-I R/IGF1R Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177680 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177680 0.1 mL
NBP177680SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP177680 Supplier Novus Biologicals Supplier No. NBP177680
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

IGF-I R/IGF1R Polyclonal specifically detects IGF-I R/IGF1R in Human, Mouse, Rat, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen IGF-I R
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:50-1:100
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. P08069
Gene Alias CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
Gene Symbols IGF1R
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human IGF1 Receptor protein (between residues 700-800) [UniProt P08069]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Nutrient Sensing in the Brain, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3480
Test Specificity Human, rat, mouse and primate. Reactivity with Equine reported via customer feedback. Predicted to react with Bovine (100%).
Target Species Human, Mouse, Rat, Equine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.