Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-I R/IGF1R Antibody (IGF1R/4667), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB179541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179541 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179541 Supplier Novus Biologicals Supplier No. NBP314094AF647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGF-I R/IGF1R Monoclonal antibody specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen IGF-I R/IGF1R
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone IGF1R/4667
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
Host Species Mouse
Immunogen Recombinant fragment (around aa550-750) of human IGF-I R/IGF1R protein (exact sequence is proprietary) (Uniprot: P08069)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3480
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.