Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-II/IGF2 Antibody (110), Biotin, Novus Biologicals™
SDP

Catalog No. NB072722 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072722 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072722 Supplier Novus Biologicals Supplier No. NBP290372B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Mouse samples. It is validated for ELISA.

Specifications

Antigen IGF-II/IGF2
Applications ELISA
Classification Monoclonal
Clone 110
Conjugate Biotin
Dilution ELISA
Formulation PBS
Gene Alias C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, PP9974, somatomedin-A
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IGF-II/IGF2 (P09535; Ala25-Glu91).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3481
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.