Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-II R/IGF2R Antibody (2G11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB300514 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300514 0.1 mL
NB300514SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300514 Supplier Novus Biologicals Supplier No. NB300514
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

IGF-II R/IGF2R Monoclonal specifically detects IGF-II R/IGF2R in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen IGF-II R/Mannose 6 Phosphate Receptor (Cation independent)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 2G11
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Flow Cytometry 1 ug per million cells, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, CyTOF-ready
Gene Accession No. P08169
Gene Alias 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR
Gene Symbols IGF2R
Host Species Mouse
Immunogen Purified bovine Mannose 6 Phosphate Receptor (Cation independent) [UniProt# P08169]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Golgi Apparatus Markers, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3482
Test Specificity Does not react with Chinese hamster ovary cell Mannose 6 Phosphate Receptor.
Target Species Human, Mouse, Rat, Bovine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.