Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-II R/IGF2R Antibody (MEM-238), FITC, Novus Biologicals™
SDP

Catalog No. NB100635991 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100635991 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100635991 Supplier Novus Biologicals Supplier No. NB10063599100TESTS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) Monoclonal specifically detects IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) in Human, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen IGF-II R/Mannose 6 Phosphate Receptor (Cation independent)
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-238
Conjugate FITC
Dilution Western Blot 1:1000, Flow Cytometry 1-2 ug/ml, Immunoprecipitation 1:10-1:500
Formulation PBS and 0.2% (w/v) high grade BSA (protease free) with 15mM Sodium Azide
Gene Alias 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR
Gene Symbols IGF2R
Host Species Mouse
Immunogen Recombinant Vaccinia virus encoding CD222.
Molecular Weight of Antigen 250 kDa
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3482
Test Specificity The antibody MEM-238 recognizes an epitope between domains 2 and 5 of CD222 (IGF2 receptor), a ubiquitously expressed 250 kDa multifunctional type I transmembrane protein. The majority of CD222 is found in the late endosomal/prelysosomal ompartment, 5-10% in the plasma membrane and the truncated (220 kDa) form of CD222 is present in human and bovine serum.
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.