Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-II R/IGF2R Antibody (MEM-238), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB500464549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500464549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500464549 Supplier Novus Biologicals Supplier No. NB500464JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGF-II R/IGF2R Monoclonal specifically detects IGF-II R/IGF2R in Human, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen IGF-IIR/Mannose6PhosphateReceptor(Cationindependent)
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-238
Conjugate Janelia Fluor 549
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR
Gene Symbols IGF2R
Host Species Mouse
Immunogen Recombinant Vaccinia virus encoding CD222.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Golgi Apparatus Markers, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3482
Test Specificity The antibody MEM-238 recognizes an epitope between domains 2 and 5 of CD222 (IGF2 receptor), a ubiquitously expressed 250 kDa multifunctional type I transmembrane protein. The majority of CD222 is found in the late endosomal/prelysosomal ompartment, 5-10% in the plasma membrane and the truncated (220 kDa) form of CD222 is present in human and bovine serum.
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.