Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-II R/IGF2R Antibody, Novus Biologicals™
SDP

Catalog No. NB10074387 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074387 Supplier Novus Biologicals Supplier No. NB10074387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) Polyclonal specifically detects IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) in Bovine, Human, Mouse, Plant, Primate samples. It is validated for Dot Blot, Immunocytochemistry/Immunofluorescence, Tissue Culture Substratum, Western Blot.

Specifications

Antigen IGF-II R/Mannose 6 Phosphate Receptor (Cation independent)
Applications Dot Blot, Immunocytochemistry, Immunofluorescence, Tissue Culture Substratum, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:3000, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Dot Blot 1:1000, Tissue Culture Substratum
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. P08169
Gene Alias 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR
Gene Symbols IGF2R
Host Species Rabbit
Immunogen 300 kDa cation-independent receptor purified from adult bovine liver tissue.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Golgi Apparatus Markers, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3482
Test Specificity CI-MPR
Target Species Bovine, Human, Mouse, Plant, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.