Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF1B Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | IGF1B |
---|---|
Concentration | 0.44 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2772
![]() ![]() |
Proteintech Group Inc
202151AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to isoform IGF-1B.
Insulin-like growth factor-1 (IGF1), is structurally and functionally related to insulin but has a much higher growth-promoting activity. A variety of cellular responses are induced by IGF1, including cell proliferation, differentiation, migration, and survival. IGF1 stimulates glucose transport in rat bone-derived osteoblastic (PyMS) cells and is effective at much lower concentrations than insulin, not only regarding glycogen and DNA synthesis but also with regards to enhancing glucose uptake. In circulation, IGFs are predomitly bound to binding proteins (IGFBPs) which prolong the half-life of the IGFs and play a role in delivering them to target tissues. Low levels of IGF1 have been linked to Alzheimer′s disease.Specifications
IGF1B | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P05019 | |
3479 | |
Peptide | |
Primary | |
-20°C | |
IGF1 |
0.44 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
IBP1, IGF I, IGF1, IGF1A, IGF1B, IGFI, Insulin like growth factor I, Mechano growth factor, MGF, Somatomedin C | |
IGF1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title