Learn More
IGF1R beta chain Mouse anti-Human, Rat, Clone: 3C6A11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | IGF1R beta chain |
---|---|
Clone | 3C6A11 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6715
![]() ![]() |
Proteintech Group Inc
662831IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody raised against a synthesized polypeptide (CGVLVLRASFDERQPYAH) corresponding to 1332-1348aa of human IGF1R protein recognizes beta chain of IGF1R.
IGF1R (Insulin-like Growth Factor1 Receptor) is a receptor tyrosine kinase that mediates actions of insulin-like growth factor 1 (IFG1). It binds IGF1 with high affinity and IGF2/insulin with lower affinity. Activated IGF1R is involved in cell growth and survival control. IGF1R is crucial for tumor transformation and survial of maligt cells. Ligand binding activates the receptor kinase and leads to receptor autophosphorylation and tyrosines phosphorylation of multiple substrates that function as signaling adapter proteins.Specifications
IGF1R beta chain | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD221, IGF I receptor, IGF1R, IGF1R-Specific, IGFIR, IGFR, JTK13 | |
IGF1R | |
IgG1 | |
Protein G | |
Antibody |
3C6A11 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
P08069, P24062 | |
25718, 3480 | |
Peptide | |
Primary | |
-20°C | |
IGF1R |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.