Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2BP1 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB362769 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362769 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362769 Supplier Novus Biologicals Supplier No. NBP179023FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGF2BP1 Polyclonal antibody specifically detects IGF2BP1 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen IGF2BP1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation 50mM Sodium Borate
Gene Alias Coding region determinant-binding protein, CRD-BP, CRDBPIGF-II mRNA-binding protein 1, IGF II mRNA binding protein 1, IMP1, IMP-1ZBP-1, insulin-like growth factor 2 mRNA binding protein 1, insulin-like growth factor 2 mRNA-binding protein 1, VICKZ family member 1, VICKZ1, ZBP1IGF2 mRNA-binding protein 1, Zip code-binding protein 1, Zipcode-binding protein 1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human Insulin-Like Growth Factor 2 mRNA Binding Protein 1 using the numbering given in entry NP_006537.3 (GeneID 10642).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10642
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.