Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF2BP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP236473
Description
IGF2BP1 Polyclonal specifically detects IGF2BP1 in Mouse, Rat samples. It is validated for Western Blot.Specifications
IGF2BP1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
IGF2BP1 | |
Synthetic peptide of Igf2bp1 Peptide sequence PQLRWEVLDSLLAQYGTVENCEQVNTESETAVVNVTYSNREQTRQAIMKL The peptide sequence for this immunogen was taken from within the described region. | |
Affinity purified | |
RUO | |
10642 | |
Mouse, Rat, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Coding region determinant-binding protein, CRD-BP, CRDBPIGF-II mRNA-binding protein 1, IGF II mRNA binding protein 1, IMP1, IMP-1ZBP-1, insulin-like growth factor 2 mRNA binding protein 1, insulin-like growth factor 2 mRNA-binding protein 1, VICKZ family member 1, VICKZ1, ZBP1IGF2 mRNA-binding protein 1, Zip code-binding protein 1, Zipcode-binding protein 1 | |
Rabbit | |
63 kDa | |
100 μL | |
Primary | |
Add 50μL of distilled water. Final anti-Igf2bp1 antibody concentration is 1mg/mL in PBS buffer with 2% sucrose. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction