Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF2BP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | IGF2BP1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23647320
![]() |
Novus Biologicals
NBP23647320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP236473
![]() |
Novus Biologicals
NBP236473 |
100 μL |
Each for $487.50
|
|
|||||
Description
IGF2BP1 Polyclonal specifically detects IGF2BP1 in Mouse, Rat samples. It is validated for Western Blot.Specifications
IGF2BP1 | |
Polyclonal | |
Rabbit | |
Coding region determinant-binding protein, CRD-BP, CRDBPIGF-II mRNA-binding protein 1, IGF II mRNA binding protein 1, IMP1, IMP-1ZBP-1, insulin-like growth factor 2 mRNA binding protein 1, insulin-like growth factor 2 mRNA-binding protein 1, VICKZ family member 1, VICKZ1, ZBP1IGF2 mRNA-binding protein 1, Zip code-binding protein 1, Zipcode-binding protein 1 | |
IGF2BP1 | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
10642 | |
IgG | |
63 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title