Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2BP3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB331435 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331435 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331435 Supplier Novus Biologicals Supplier No. NBP297506JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGF2BP3 Polyclonal antibody specifically detects IGF2BP3 in Human samples. It is validated for Western Blot

Specifications

Antigen IGF2BP3
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CT98, hKOC, IGF II mRNA binding protein 3, IGF2 mRNA-binding protein 3, IGF-II mRNA-binding protein 3, IMP-3KH domain-containing protein overexpressed in cancer, IMP3VICKZ family member 3, insulin-like growth factor 2 mRNA binding protein 3, insulin-like growth factor 2 mRNA-binding protein 3, KH domain containing protein overexpressed in cancer, KOC1DKFZp686F1078, VICKZ3cancer/testis antigen 98
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human IGF2BP3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10643
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.