Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2BP3 Rabbit anti-Human, Clone: 8K4H5, Novus Biologicals™
SDP

Catalog No. NB167502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167502 20 μg
NB167501 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167502 Supplier Novus Biologicals Supplier No. NBP31620520UL

Rabbit Monoclonal Antibody

IGF2BP3 Monoclonal antibody specifically detects IGF2BP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen IGF2BP3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8K4H5
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CT98, hKOC, IGF II mRNA binding protein 3, IGF2 mRNA-binding protein 3, IGF-II mRNA-binding protein 3, IMP-3KH domain-containing protein overexpressed in cancer, IMP3VICKZ family member 3, insulin-like growth factor 2 mRNA binding protein 3, insulin-like growth factor 2 mRNA-binding protein 3, KH domain containing protein overexpressed in cancer, KOC1DKFZp686F1078, VICKZ3cancer/testis antigen 98
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 2-194 of human IGF2BP3 (O00425).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10643
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.