Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-1 Antibody (OTI6C7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289198 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289198 Supplier Novus Biologicals Supplier No. NBP271011APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGFBP-1 Monoclonal antibody specifically detects IGFBP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen IGFBP-1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6C7
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 26-259 of human IGFBP1(NP_000587) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Diabetes Research, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3484
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.