Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-1 Antibody (OTI6C7), HRP, Novus Biologicals™
SDP

Catalog No. NBP271011H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271011H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271011H Supplier Novus Biologicals Supplier No. NBP271011H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGFBP-1 Monoclonal specifically detects IGFBP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen IGFBP-1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6C7
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP
Gene Symbols IGFBP1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 26-259 of human IGFBP-1 (NP_000587) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3484.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.