Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-2 Antibody (CL13565), Novus Biologicals™
SDP

Catalog No. NB306031 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306031 25 μL
NB306032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB306031 Supplier Novus Biologicals Supplier No. NBP32456925UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGFBP-2 Monoclonal antibody specifically detects IGFBP-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen IGFBP-2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL13565
Conjugate Unconjugated
Dilution Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2
Host Species Mouse
Immunogen This antibody was designed to target the recombinant protein IGFBP-2, with the precise immunogen sequence being withheld for proprietary reasons.
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytokines and Growth Factors, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3485
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.