Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-3 Antibody (MM0342-6U23), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB360995 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360995 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360995 Supplier Novus Biologicals Supplier No. NBP212364APC

Mouse Monoclonal Antibody

IGFBP-3 Monoclonal antibody specifically detects IGFBP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen IGFBP-3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0342-6U23
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Host Species Mouse
Immunogen Human recombinant IGFBP-3
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3486
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.