Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-3 Antibody (MM0342-6U23) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019193 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019193 0.025 mg
NBP212364 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019193 Supplier Novus Biologicals Supplier No. NBP2123640.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGFBP-3 Monoclonal specifically detects IGFBP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IGFBP-3
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MM0342-6U23
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Immunohistochemistry 1:20-200, Immunohistochemistry-Paraffin 1:20-200
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. CAA46087
Gene Alias acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Gene Symbols IGFBP3
Host Species Mouse
Immunogen Human IGFBP3
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3486
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.