Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-3 Antibody (RM0096-11B16), Biotin, Novus Biologicals™
SDP

Catalog No. NBP122452B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122452B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122452B Supplier Novus Biologicals Supplier No. NBP122452B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IGFBP-3 Monoclonal specifically detects IGFBP-3 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IGFBP-3
Applications Western Blot
Classification Monoclonal
Clone RM0096-11B16
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Gene Symbols IGFBP3
Host Species Rat
Immunogen Mouse recombinant protein of IGFBP-3
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Biologically Active Proteins, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3486
Test Specificity This antibody detects mouse IGFBP-3 in Western blotting. No cross activity to other mouse IGFBPs.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.