Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-4 Antibody (IBP180), Novus Biologicals™
SDP

Catalog No. NBP232771 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232771 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232771 Supplier Novus Biologicals Supplier No. NBP232771
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGFBP-4 Monoclonal antibody specifically detects IGFBP-4 in Human samples. It is validated for ELISA.

Specifications

Antigen IGFBP-4
Applications ELISA
Classification Monoclonal
Clone IBP180
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000 Sandwich ELISA 1:100-1:2000
Gene Alias BP-4, HT29-IGFBP, IBP4insulin-like growth factor-binding protein 4, IGF-binding protein 4, IGFBP-4IBP-4, insulin-like growth factor binding protein 4
Gene Symbols IGFBP4
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant human IGFBP-4 expressed in mammalian cell line.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3487
Test Specificity Human N-terminal IGFBP-4 fragment (NT-IGFBP-4)
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.