Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-5 Antibody (MM0344-8D21), FITC, Novus Biologicals™
SDP

Catalog No. NBP212366F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212366F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212366F Supplier Novus Biologicals Supplier No. NBP212366F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGFBP-5 Monoclonal specifically detects IGFBP-5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IGFBP-5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0344-8D21
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias IBP5IBP-5, IGF-binding protein 5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5
Gene Symbols IGFBP5
Host Species Mouse
Immunogen Human recombinant IGFBP-5
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3488
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.