Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-5 Antibody (RM0097-12C23) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018433 0.025 mg
NBP122453 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018433 Supplier Novus Biologicals Supplier No. NBP1224530.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IGFBP-5 Monoclonal specifically detects IGFBP-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IGFBP-5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RM0097-12C23
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q07079
Gene Alias IBP5IBP-5, IGF-binding protein 5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5
Gene Symbols IGFBP5
Host Species Rat
Immunogen Mouse recombinant protein of IGFBP-5
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3488
Test Specificity This antibody detects mouse IGFBP-5 in Western blotting. No cross activity to other mouse IGFBPs.
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.