Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-5 Antibody (RM0097-12C23) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | IGFBP-5 |
---|---|
Clone | RM0097-12C23 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Description
IGFBP-5 Monoclonal specifically detects IGFBP-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IGFBP-5 | |
LYOPH | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
IBP5IBP-5, IGF-binding protein 5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5 | |
IGFBP5 | |
IgG2 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
RM0097-12C23 | |
Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Q07079 | |
3488 | |
Mouse recombinant protein of IGFBP-5 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title