Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-rp1/IGFBP-7 Antibody (115), PerCP, Novus Biologicals™
SDP

Catalog No. NB316661 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316661 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316661 Supplier Novus Biologicals Supplier No. NBP290176PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IGFBP-rp1/IGFBP-7 Monoclonal antibody specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen IGFBP-rp1/IGFBP-7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 115
Conjugate PerCP
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP-rp1/IGFBP-7 (Uniprot#: Q16270; Met1-Leu282).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 3490
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.