Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-rP10/KAZALD1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB318281 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318281 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318281 Supplier Novus Biologicals Supplier No. NBP298914MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGFBP-rP10/KAZALD1 Polyclonal antibody specifically detects IGFBP-rP10/KAZALD1 in Human samples. It is validated for Western Blot

Specifications

Antigen IGFBP-rP10/KAZALD1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias bone- and odontoblast-expressed gene 1, BONO1, FKSG40, FLJ24094, IGFBP-related protein 10, IGFBP-rP10, Kazal-type serine peptidase inhibitor domain 1, Kazal-type serine protease inhibitor domain 1, kazal-type serine protease inhibitor domain-containing protein 1, novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein, UNQ2945/PRO21184
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IGFBP-rP10/KAZALD1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81621
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.