Learn More
Invitrogen™ IGFL3 Polyclonal Antibody

Description
Immunogen sequence: TTDAPVGSGL WLCQPTPRCG NKIYNPSEQC CYDDAILSLK ETRRCGSTCT FWPCFELCCP ESFGPQQKFL VKLRVLGMKS QCHLSPISRS CTRNRRHVLY P.
Specifications
Specifications
| Antigen | IGFL3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.24 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | IGFL3 |
| Gene Accession No. | Q6B9Z0 |
| Gene Alias | Gm580; IGF; IGF like family member 3; Igfl; Igfl3; IGF-like family member 3; Insulin growth factor-like family member; insulin growth factor-like family member 3; Insulin like growth factor; mIgfl; Rprc483; UNQ483; UNQ483/PRO982 |
| Gene Symbols | IGFL3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.