Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFLR1/TMEM149 Antibody (905338), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB310897 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310897 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310897 Supplier Novus Biologicals Supplier No. FAB8296PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGFLR1/TMEM149 Monoclonal specifically detects IGFLR1/TMEM149 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IGFLR1/TMEM149
Applications Flow Cytometry
Classification Monoclonal
Clone 905338
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias FLJ22573, transmembrane protein 149, U2 small nuclear RNA auxiliary factor 1-like 4, U2(RNU2) small nuclear RNA auxiliary factor 1-like 4, U2AF1L4
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human IGFLR1, Met1-Pro163, Accession # Q9H665
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79713
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.