Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG F(ab)2 Goat anti-Mouse, Allophycocyanin, Polyclonal, Seconday Antibody, Novus Biologicals™
SDP

Catalog No. NB008596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008596 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008596 Supplier Novus Biologicals Supplier No. NBP261929AP0.5ML

Goat Polyclonal Antibody

IgG F(ab')2 Polyclonal specifically detects IgG F(ab')2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IgG F(ab)2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate APC
Dilution Western Blot 1:2500-1:25000, ELISA 1:2500-1:25000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 50 mM HEPES pH 7.1, 0.1 M NaCl, 1 mM MgCl2, 0.1 mM ZnCl2 containing 0.2% BSA.
Host Species Goat
Immunogen Mouse IgG-F(ab')2 Fragment
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Antiserum was solid phase adsorbed to ensure class specificity. Antiserum was cross adsorbed using human and rat immunosorbents to remove cross reactive antibodies. The antibody to mouse F(ab')2 was isolated by affinity chromatography using antigen coupled to agarose beads. F(ab')2 fragments were generated using a pepsin digestion. Fc fragments and whole IgG molecules have been removed. Fragments were conjugated to alkaline phosphatase. Antibody concentration was determined by extinction coefficient: absorbance at 280 nm of 1.4 equals 1.0 mg of IgG. By immunoelectrophoresis and ELISA this antibody reacts specifically with mouse F(ab')2. No antibody was detected against non-immunoglobulin serum proteins. Less than 1% cross reactivity to human and rat F(ab')2 was detected. This antibody may cross react with F(ab')2 from other species.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.