Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Fc Antibody (RMG06) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB301087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301087 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301087 Supplier Novus Biologicals Supplier No. NBP325962
Add to Cart
Add to Cart

Goat Monoclonal Antibody

IgG Fc Monoclonal antibody specifically detects IgG Fc in Mouse samples. It is validated for ELISA

Specifications

Antigen IgG Fc
Applications ELISA
Classification Monoclonal
Clone RMG06
Conjugate Unconjugated
Dilution ELISA : 0.05ug/ml - 1ug/ml
Formulation PBS
Host Species Goat
Immunogen Mouse IgG
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.