Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Light Chain Antibody (IVA285-1), FITC, Novus Biologicals™
SDP

Catalog No. NB002161 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002161 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002161 Supplier Novus Biologicals Supplier No. NBP262190F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG Light Chain Monoclonal specifically detects IgG Light Chain in Bovine, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen IgG Light Chain
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone IVA285-1
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Fraction of Bovine immunoglobulin.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Bovine, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.