Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Mouse anti-Human, Clone: SPM566, Novus Biologicals™
SDP

Catalog No. NB34381200U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB34381200U 200 μg
NB3438120UG 20 μg
NB34381100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB34381200U Supplier Novus Biologicals Supplier No. NBP234381200UG

Mouse Monoclonal Antibody

ELISA (For coating, order NBP2-34428 (BSA free version of NBP2-34381))

Specifications

Antigen IgG
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM556
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Alias immunoglobulin heavy constant gamma 1 (G1m marker)
Gene Symbols IGHG1
Host Species Mouse
Immunogen Purified human Ig Gamma Chain
Molecular Weight of Antigen 75 kDa
Purification Method Protein A purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Test Specificity Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes.
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2a κ
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.