Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (Whole Ab), Sheep, Biotin, Polyclonal, Cytiva

Catalog No. 45200581 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-200-581 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-200-581 Supplier Cytiva Supplier No. RPN10012ML
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

  • IgG (Whole Ab) Polyclonal antibody specifically detects Antigen in Ms samples. It is validated for ELISA, Immunocytochemistry, Western Blotting
  • Soluble in cold water and hot water
  • Shelf Life: 12 months from the date of manufacture

Order Info

Highly species-specific

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (Whole Ab)
Applications Immunocytochemistry, Protein Blot
Classification Polyclonal
Conjugate Biotin
Host Species Sheep
Immunogen Mouse Ig, whole molecule
Purification Method Affinity Purified
Quantity 2 mL
Primary or Secondary Primary
Includes Phosphate buffered saline (sodium phosphate 0.1M, NaCl 0.1M) pH 7.5, containing 1% (w/v) bovine serum albumin and 0.05% (w/v) sodium azide
Target Species Mouse
Content And Storage 2 to 8°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.