Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Antibody (RM117), Biotin, Novus Biologicals™
SDP

Catalog No. NB002146 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002146 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002146 Supplier Novus Biologicals Supplier No. NBP262028B0.05MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgG1 Monoclonal specifically detects IgG1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen IgG1
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone RM117
Concentration 1 mg/mL
Conjugate Biotin
Dilution Flow Cytometry, ELISA 0.05 - 0.2 μg/mL, Immunohistochemistry 0.5 - 2 μg/mL, Immunocytochemistry 0.5 - 2 μg/mL
Formulation PBS, 50% Glycerol, 1% BSA
Host Species Rabbit
Immunogen Peptide corresponding to the hinge region of Human IgG1
Quantity 0.05 mg
Test Specificity This antibody reacts to the heavy chain of human IgG1. It does not cross react to any other IgG subclasses (IgG2, IgG3, or IgG4), and shows no cross reactivity to IgM, IgA, IgD, IgE. This antibody does not react to monkey (Cyno or Rhesus) IgG, mouse IgG, rat IgG, or goat IgG.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.