Learn More
Description
Specifications
Specifications
| Antigen | IgG1 Fc |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 43D17 |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot, ELISA |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | IgG1, Igh-4, RGD1359539, VH7183 |
| Host Species | Mouse |
| Immunogen | Human recombinant IgG1 Fc |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
