Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Fc Antibody (43D17), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB360814 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360814 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360814 Supplier Novus Biologicals Supplier No. NBP222028APC

Mouse Monoclonal Antibody

IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen IgG1 Fc
Applications Western Blot, ELISA
Classification Monoclonal
Clone 43D17
Conjugate APC
Dilution Western Blot, ELISA
Formulation PBS
Host Species Mouse
Immunogen Human recombinant IgG1 Fc
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.