Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2 Mouse anti-Human, PE, Clone: HP6002 (also known as AB4-DH4), BD Pharmingen™
Mouse Monoclonal Antibody
$158.00 - $355.50
Specifications
Antigen | IgG2 |
---|---|
Clone | HP6002 (also known as AB4-DH4) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
IgG2 Monoclonal antibody specifically detects IgG2 in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
IgG2 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
IGHG2; Immunoglobulin heavy constant gamma 2 | |
Purified Human IgG2 Myeloma Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
HP6002 (also known as AB4-DH4) | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
3501 | |
IgG1 κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title