Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2b Rat anti-Mouse, Clone: MG2b, Novus Biologicals™
SDP

Catalog No. NB171954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1000 μg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171954 1000 μg
NB171955 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171954 Supplier Novus Biologicals Supplier No. NBP3183071000UG
This item is not returnable. View return policy

Rat Monoclonal Antibody

IgG2b Monoclonal antibody specifically detects IgG2b in Mouse samples. It is validated for ELISA

Specifications

Antigen IgG2b
Applications ELISA
Classification Monoclonal
Clone MG2b
Concentration 0.5 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered PBS
Host Species Rat
Immunogen Mouse IgG
Purification Method Ion exchange chromatography
Quantity 1000 μg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.