Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2c Rat anti-Mouse, Clone: MTG2c, Novus Biologicals™
SDP

Catalog No. NB171957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1000 μg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171957 250 μg
NB171956 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171957 Supplier Novus Biologicals Supplier No. NBP318308250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IgG2c Monoclonal antibody specifically detects IgG2c in Mouse samples. It is validated for ELISA

Specifications

Antigen IgG2c
Applications ELISA
Classification Monoclonal
Clone MTG2c
Concentration 0.5 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered PBS
Host Species Rat
Immunogen Mouse IgG
Purification Method Protein G purified
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.