Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG3 Antibody (RM218) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB301095 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301095 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301095 Supplier Novus Biologicals Supplier No. NBP325954
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgG3 Monoclonal antibody specifically detects IgG3 in Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen IgG3
Applications Western Blot, ELISA
Classification Monoclonal
Clone RM218
Conjugate Unconjugated
Dilution Western Blot : 0.1 ug/ml - 0.5 ug/ml, ELISA : 0.005 ug/ml-0.2 ug/ml
Formulation PBS
Gene Alias DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Host Species Rabbit
Immunogen Mouse IgG3
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3502
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.