Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgJ Antibody (027), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB070402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070402 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070402 Supplier Novus Biologicals Supplier No. NBP290155UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgJ Monoclonal specifically detects IgJ in Human samples. It is validated for ELISA.

Specifications

Antigen IgJ
Applications ELISA
Classification Monoclonal
Clone 27
Conjugate DyLight 350
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias IGCJJCH, immunoglobulin J chain, immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mupolypeptides
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IgJ (NP_653247.1; Gln23-Asp159).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3512
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.